AC-heat controller with solid-state relay
Representative image only.
Item # EW-03052-65
  • 6 Month Product / 12 Month Parts and Labor Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Input (VAC)115
  • Output (VAC)115
  • DescriptionAC-heat controller with solid-state relay
  • Warranty6 Month Product / 12 Month Parts and Labor Warranty
Key Features
  • 6 Month Product / 12 Month Parts and Labor Warranty
More About this Item
Maintain precise thermail control over your resistive heating devices. These heat controllers proportion the amount of delivered line voltage up to 1500 watts. Use the stepless, adjustment knob to easily change the ratio of "ON" to "OFF" power time.
Choose from two models-controller 03052-65 features a solid-state relay which eliminates arcing switch contacts. Both models include a 6-ft, three-wire cord (UL-approved) and plug for 115 VAC operation. Controllers measure 3-1/4"W x 7"H x 4"D.