3M 8210 Particulate Respirators, N95, Standard Particulate, Box of 20
Representative image only.
3M Mfr # 8210 – Item # EW-40111-02
Industry standard for protection against solid and liquid particles
  • NIOSH approved
PRODUCT OPTIONS
$31.80USD / Box of 20
BUY MORE & SAVE
  • 1-9$31.80USD / Box of 20
  • 10+$28.62USD / Box of 20
SPECS & DESCRIPTION
Specifications & Description
  • RatingN95
  • ComplianceNIOSH
  • ColorWhite
  • 3M Item Number8210
  • DescriptionParticulate Respirators, N95, Standard Particulate, Box of 20
Key Features
  • NIOSH approved
More About this Item
These lightweight respirator masks offer comfort and adjustable straps for a snug fit.
Standard N95 is the economical choice for grinding, sanding, sweeping, bagging, and other work in dusty environments.