3M 330-3001 Standard Earmuff, 20 dB NRR; 1/Pk
Representative image only.
3M Mfr # 330-3001 – Item # EW-48492-00
Universal headband fits any head size
SPECS & DESCRIPTION
Specifications & Description
  • NRR (dB)20
  • ComplianceANSI S3.19-1974
  • DescriptionStandard Earmuff, 20 dB NRR; 1/Pk
More About this Item
Three-position muff is made of durable, lightweight plastic for use in tough environments. Soft, foam-filled cushions and large earcup openings give you extra protection and are comfortable to wear. Pivoting earcup connection maintains seal and alignment.