3M 11374-00000 Eyewear, Silver/Black frame, Clear lens Reader, 1.5 Diopter
Representative image only.
3M Mfr # 11374-00000-20 – Item # EW-86472-63
Three lens angles for a custom fit
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
PRODUCT OPTIONS
$37.40USD / Each
BUY MORE & SAVE
  • 1-9$37.40USD / Each
  • 10+$33.66USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorBlack/Silver
  • Lens ColorClear
  • ComplianceANSI Z87.1-2003; CSA Z94.3
  • Magnification+1.5
  • DescriptionEyewear, Silver/Black frame, Clear lens Reader, 1.5 Diopter
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
High-impact polycarbonate lens blocks 99.9% UV and features antifog coating. Glasses feature a soft, universal fit nosebridge and soft temple provide comfort for long periods of wear. Temple is adjustable to three lengths. Reader magnifiers available in 1.5, 2.0, and 2.5 power
Compliance: ANSI Z87.1-2003, CSA Z94.3