Cole-Parmer 316 Stainless Steel Shaft, 3/8" dia x 12"L (accepts 5/16" bore dia propeller)
Representative image only.
Cole-Parmer Item # EW-04552-25
Keep your overhead mixer running smoothly
  • 2 year warranty
PRODUCT OPTIONS
$62.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Description316 Stainless Steel Shaft, 3/8\" dia x 12\"L (accepts 5/16\" bore dia propeller)
  • Warranty2 year
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Update clamps, mounts, shafts and propellers to accommodate new protocols. These replacement parts will keep your overhead mixer running smoothly.