Home  >  Bromoq to Buffer >
  

Buffer Solution (Acetate), pH 4.0-4.5 (Certified) (20 liter) (SB10320)

YO-88084-02
Loading price...

Usually ships in 14 days.
Qty


    Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

    This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
    Product Type Chemical Buffers
    Purity grade Certified
    CAS Number 6131-90-4 (Sodium Acetate, Trihydrate), 64-19-7 (Acetic Acid, Glacial)
    Form Liquid
    Packaging PolyPac
    Storage symbol Gray
    Quantity (mL) 20000
    Manufacturer number SB10320
    Quantity 20 liter





    Technical Resources
     
    Related Products for Buffer Solution (Acetate), pH 4.0-4.5 (Certified) (20 liter) (SB10320)
    Image
      Product Detail
    SKU

    Buffer Solution, pH 10.00 Color-Coded Blue (Certified) (20 liter) (YO-88084-21)

    Category: Bromoq to Buffer
    Description: Buffer Solution, pH 10.00 Color-coded Blue
    Brand: Cole-Parmer


    Buffer Solution, pH 9.00 (Certified) (20 liter) (SB11420)

    Category: Bromoq to Buffer
    Description: Buffer Solution, pH 9.00
    Brand: Cole-Parmer


    Buffer Solution, pH 6.00 (Certified) (20 liter) (SB10420)

    Category: Bromoq to Buffer
    Description: Buffer Solution, pH 6.00
    Brand: Cole-Parmer


    Buffer Solution, pH 8.00 (Certified) (20 liter) (SB11220)

    Category: Bromoq to Buffer
    Description: Buffer Solution, pH 8.00
    Brand: Cole-Parmer


    Buffer Solution, pH 10.00 (Certified) (20 liter) (SB11620)

    Category: Bromoq to Buffer
    Description: Buffer Solution, pH 10.00
    Brand: Cole-Parmer


    Buffer Solution, pH 2.00 (Certified) (20 liter) (SB9620)

    Category: Bromoq to Buffer
    Description: Buffer Solution, pH 2.00
    Brand: Cole-Parmer


    Buffer Solution, pH 3.00 (Certified) (20 liter) (SB9720)

    Category: Bromoq to Buffer
    Description: Buffer Solution, pH 3.00
    Brand: Cole-Parmer


    Buffer Solution, pH 4.00 (Certified) (20 liter) (SB9820)

    Category: Bromoq to Buffer
    Description: Buffer Solution, pH 4.00
    Brand: Cole-Parmer


    Hydrochloric Acid Solution (0.2N (N/5)/Certified) (20 liter) (SA5220)

    Category: Homatro to Hydroq
    Description: Hydrochloric Acid Solution
    Brand: Cole-Parmer


    Cole-Parmer NIST-Traceable pH Buffer Standard Solution, pH 4; 500 mL

    Category: Buffers and Solutions for pH/Ion Measurement
    Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
    Brand: Cole-Parmer


    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers
    Product Reviews

    Buffer Solution (Acetate), pH 4.0-4.5 (Certified) (20 liter) (SB10320)
    Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
    Popularity |  Favorites |  Title |  Price |  Part Number |  Category
    Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

    Previous: Buffer Salt, pH 9.18 (Certified) (12 packets per pack)  |  Next: Buffer Solution, pH 10.00 (Certified) (20 liter) (SB11620)